Corning® HYPERStack® Nest
Fournisseur: Corning
10047
CORN10047EA 252 EUR InStock
CORN10047
Corning® HYPERStack® Nest
Portoirs Portoirs pour flacons
The HYPERStack® Nest accessory is designed for use with the Corning® HYPERStack® 36-layer cell culture vessel.
  • Lightweight design: 2 lbs. (950 g)
  • Simplify process consistency by consistently placing the vessel in the correct position to facilitate optimal fill and empty processes
  • Designed to reduce the chance of pressurizing vessels and wetting filters
  • Pivot angle precisely engineered in the Nest reduces lifting by the user and freeing hands to operate tubing clamps
  • Made of stainless steel, the Nest can be sterilized and autoclaved
  • Small footprint fits
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité