384-well microplate black/clear, round bottom, ultra-low attachment, spheroid, Corning®
Fournisseur: Corning
3830BC
CORN3830BCEA 2490 EUR InStock
CORN3830BC
384-well microplate black/clear, round bottom, ultra-low attachment, spheroid, Corning®
Microplaques Microplaques pour culture cellulaire
Ultra-Low Attachment microplates feature a covalently bound hydrogel layer that effectively inhibits cellular attachment.
  • Sterilised by gamma irradiation
  • Opaque walls to prevent well-to-well crosstalk
  • Surface is noncytotoxic, biologically inert and nondegradable
  • With Generic Bar codes, and lid
Surface minimises protein absorption, enzyme activation and cellular activation; Black wall microplates have low background fluorescence, minimal light scatter and reduced crosstalk.
Can be used for both top and bottom reading instruments. Novel well geometry aids formation of spheroids in center of well. Optically clear round bottom with black opaque microplate body
Surface treatment: Ultra-low attachment.
Conditionnement: Bulk packed 10 per bag.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité