1536-well spheroid microplates, Corning®
Fournisseur: Corning
4637
CORN4637EA 644 EUR InStock
CORN4637
1536-well spheroid microplates, Corning®
Microplaques Microplaques pour culture cellulaire
With their proprietary design, these microplates are ideal for generating and analyzing 3D multicellular spheroids in the same microplate. The Ultra-Low Attachment (ULA) surface enables uniform and reproducible 3D multicellular spheroid formation. The black opaque microplate body shields each optically clear, round bottom well from well-to-well cross-talk.
  • 536-well format allows for high throughput 3D cell culture
  • Optically clear round bottom with black opaque microplate body
  • Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
  • Novel well geometry aids in the generation of uniform, single spheroids across all wells, which enables automated visualization
  • Unique design shields each well to minimize well-to-well cross-talk
You can culture and assay spheroids in the same microplate, without the need for transfer to a new microplate. Compatible with the GNF ultra high throughput system.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité