AseptiQuik® X connectors
Fournisseur: CPC COLDER
AQX33024HT AQX44024HT
CPCCAQX33024HTEA 231 EUR InStock
CPCCAQX33024HT CPCCAQX44024HT
AseptiQuik® X connectors
Raccords et connecteurs
AseptiQuik® X large format 1" connectors provide quick and easy sterile connections for high flow applications, even in non-sterile environments. The 'Twist-Pull-Twist' design enables users to quickly transfer large volumes of media easily with less risk of operator error.
  • Operating temperature: 4 to 40 °C
  • Ensure simultaneous and secure removal of both membranes
  • Audible confirmation of completed assembly steps
  • Open access through multiple supply chain partners
  • Secures final connection preventing disassembly
Biopharmaceutical manufacturers can now make 1" sterile connections with the quality and market availability they expect from the leader in single-use connection technology.
Certifications: USP Class VI, ADCF Certified.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité