Gear loops, Protecta®, 3M™
Fournisseur: CAPITAL SAFETY DEUTSCHLAND
1150492
CPSD1150492EA 0 EUR InStock
CPSD1150492
Gear loops, Protecta®, 3M™
Protections anti-chutes Câbles de sécurité
PROTECTA® Gear loops are a part of the fall arrest harness accessory options. They provide the opportunity to add extra tool carrying convenience to a harness.
  • Tools can be secured while working at heights
  • Keep people and property safe anywhere below an elevated worksite
  • Suitable for use with 3M™ PROTECTA® Comfort belt harness models only
These are part of a range of optional extras, which are available to upgrade the comfort and convenience of a PROTECTA® Comfort belt style fall arrest harness.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité