Quick-wrap tape II, DBI-SALA®
Fournisseur: CAPITAL SAFETY DEUTSCHLAND
1500175
CPSD1500175EA 0 EUR InStock
CPSD1500175
Quick-wrap tape II, DBI-SALA®
Rubans adhésifs
DBI-SALA® Quick-Wrap tape II helps to deliver long-lasting tool attachment performance while requiring up to 40% less tape compared to other Quick-Wrap tapes.
  • Conforms to the shape of tool handles
  • Adheres on contact to most clean tools and can be repositioned during installation
  • Enhanced stretching results in a thin, low-profile installation
  • Ready for use upon installation
  • No waiting period required
  • Connection points on tools up to 0,9 kg
The material and adhesive technology helps provide good tear resistance while you work, the high-elongation properties result in more wraps with the same length of tape.
Length: 2,7 m
Colour: Yellow
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité