Fixed beam anchor, DBI-SALA, 3M™
Fournisseur: CAPITAL SAFETY
2108413
CPSD2108413EA 0 EUR InStock
CPSD2108413
Fixed beam anchor, DBI-SALA, 3M™
Protections anti-chutes Cordes de sécurité
The DBI-SALA® fixed beam anchor fits 63 to 457 mm wide i-beams up to 38 mm thick. It installs in seconds to the beam structure in any orientation with final tightening using a built-in adjustment handle.
  • Attach in any orientation or direction
  • Compact and lightweight design
  • High strength steel and aluminum construction
  • Swiveling D-ring tie-off point
  • Detent style attachment with built-in tightening handle
This anchorage connector is made from top quality materials guaranteeing high strength and durability to stand up to the toughest environments.
Capacity: 140 kg
Certifications: EN 795, Class B
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité