Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Confined space high capacity short reach davit system, DBI-SALA, 3M™

Fournisseur: CAPITAL SAFETY DEUTSCHLAND
8000118
CPSD8000118EA 0 EUR InStock
CPSD8000118
Confined space high capacity short reach davit system, DBI-SALA, 3M™
Protections anti-chutes Cordes de sécurité
The DBI-SALA® Confined space HC (high capacity) short reach davit system is constructed of powder-coated aluminium, and features a modular design for easy transport and storage. The system is adjustable, providing flexibility during installation.

  • Modular design can be separated for easy transportation and storage
  • Provides a complete confined space rescue and work support system
  • System comes with 3-piece base, 84cm mast extension, and short reach davit arm
  • Adjustable offset from 36 to 71cm (14 to 28")
  • Adjustable vertical height from 180 to 228cm (71 to 90")
  • 180° davit mast rotation
  • Adjustable width base design with integrated levelling feet and indicator

This features a modular design, allowing it to be separated into multiple pieces for easy transport and storage. Davit arm height and reach are both adjustable, and securely locked using detent pins which are attached with retracting pin lanyards that help prevent breaks and snags. This system is constructed primarily of custom-extruded aluminium, and finished in a mixture of hi-visibility green and gloss-black powder coatings. The new design makes the davit height adjustment process two times faster than previous models. System width is adjustable and locked using detent pins providing flexibility during installation. The base is equipped with integrated alignment features that enable easy assembly, and integrated levelling feet and level indicators which help ensure proper setup.

Compared to previous models, a larger safe working coverage area is allowed with this base. Furthermore, accompanying davit arms can be used in all pin positions, and with equipment (SRL/Winch) front or back mounted. Four independent anchorage locations are available. Multiple user fall arrest is supported for up to two users weighing 140kg each. Material handling capabilities up to 205 kg are made possible by meeting the requirements of the machinery directive for manually operated lifting 2006/42EC.

Weight: 59,06 kg.

Certifications: Certified to EN 795:2012, CEN/TS 16415:2013, EN 1496:2012, and meets the requirements of machinery directive 2006/42EC for material handling.

Commander maintenant
 

COMMANDE

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité