Supelco® TraceCERT® Organic Reference Standard, Cyantraniliprole
Fournisseur: Merck
CRM01303-25MG
CRM01303-25MGEA 125 EUR InStock
CRM01303-25MG
Supelco® TraceCERT® Organic Reference Standard, Cyantraniliprole
Étalons Étalons organiques
It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Cyantraniliprole certified reference material (CRM) may also find following use:
  • Study of DNA damage, oxidative stress, and biological toxicity caused in earthworm by cyantraniliprole
  • Development of an isotope-labelled internal standard based method using ultra-high-performance liquid chromatography-tandem mass spectrometry for the analysis of chlorantraniliprole and cyantraniliprole in fruits, vegetables, and cereals
  • Simultaneous determination of 8 new generation amide insecticides by ultrahigh performance liquid chromatography-tandem mass spectrometry in complex food matrices
  • Estimation of five diamide insecticides using carbon nanotube multiplug filter for sample clean-up and ultrahigh-performance liquid chromatography-tandem mass spectrometry in food matrices
  • Simultaneous analysis of five diamide insecticides in edible mushrooms by high-performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS) and a modified QuEChERS procedure for extraction
Attention: P273 - P391 - P501
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité