Supelco® TraceCERT® Organic Reference Standard, Fluopyram
Fournisseur: Merck
CRM01345-25MG
CRM01345-25MGEA 92.4 EUR InStock
CRM01345-25MG
Supelco® TraceCERT® Organic Reference Standard, Fluopyram
Étalons Étalons organiques
It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Fluopyram CRM may also find its use as described below:
  • Method development for analyzing the succinate-dehydrogenase-inhibitor fungicide residue levels in vegetable and fruit samples by liquid chromatography-tandem mass spectrometry
  • Development and validation of monoclonal antibody-based immunoassays to analyze fluopyram residues in food samples
  • Simultaneous estimation of fluopyram and tebuconazole in watermelon and soil samples using QuEChERS and GC-MS based method
  • Determination of fluopyram flupyradifurone and indaziflam in date fruit pistachio and soil samples by ultra-high performance liquid chromatography with tandem mass spectrometry(UHPLC-MS)
  • Develop and validate a method based on QuEChERS and UHPLC-Orbitrap-MS to determine fluopyram and its metabolites in cherry tomato and cucumber
Attention: P273 - P391 - P501
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité