Supelco® TraceCERT® Organic Reference Standard, Tebufenozide
Fournisseur: Merck
CRM01373-50MG
CRM01373-50MGEA 101 EUR InStock
CRM01373-50MG
Supelco® TraceCERT® Organic Reference Standard, Tebufenozide
Étalons Étalons organiques
It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Tebufenozide certified reference material (CRM) may also find the following uses:
  • Analysis of trifloxystrobin tebufenozide and halofenozide by Micellar electrokinetic capillary chromatography (MEKC) in tomato celery and apple juice samples
  • Measurement of the residue levels of tebufenozide in cabbage and soil by reversed-phase high-performance liquid chromatography (HPLC)
  • Determination of four diacylhydrazine insecticide residues in apple grape cucumber tomato and spinach samples by QuEChERS method using ultra-performance liquid chromatography coupled with tandem mass spectrometry
  • Development and validation of a multi-residual analytical method based on SPE followed by LC-MS for the simultaneous estimation of 14 pesticides in surface and groundwaters
  • Multi-residue analysis of 200 pesticides and pesticide metabolites in honeybees by gas and liquid chromatography coupled with tandem mass spectrometry
Certifications: It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Tebufenozide certified reference material (CRM) may also find the following uses:
  • Analysis of trifloxystrobin tebufenozide and halofenozide by Micellar electrokinetic capillary chromatography (MEKC) in tomato celery and apple juice samples
  • Measurement of the residue levels of tebufenozide in cabbage and soil by reversed-phase high-performance liquid chromatography (HPLC)
  • Determination of four diacylhydrazine insecticide residues in apple grape cucumber tomato and spinach samples by QuEChERS method using ultra-performance liquid chromatography coupled with tandem mass spectrometry
  • Development and validation of a multi-residual analytical method based on SPE followed by LC-MS for the simultaneous estimation of 14 pesticides in surface and groundwaters
  • Multi-residue analysis of 200 pesticides and pesticide metabolites in honeybees by gas and liquid chromatography coupled with tandem mass spectrometry
Attention: P273 - P391 - P501
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité