Supelco® TraceCERT® Organic Reference Standard, Teflubenzuron
Fournisseur: Merck
CRM01399-100MG
CRM01399-100MGEA 115 EUR InStock
CRM01399-100MG
Supelco® TraceCERT® Organic Reference Standard, Teflubenzuron
Étalons Étalons organiques
It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. The teflubenzuron certified reference material (CRM) may also find use as given below:
  • Determination of 9 benzoylurea insecticides in fruit vegetable cereal and animal product samples by pressurised liquid extraction and LC-MS/MS
  • Simultaneous analysis of lufenuron bifenthrin and teflubenzuron residues in coconut water by matrix solid-phase dispersion extraction combined with reversed-phase high-performance liquid chromatography
  • Development of a high performance liquid chromatography-triple quadrupole mass spectrometry (HPLC-MS/MS) based method for the simultaneous estimation of 7 benzoylurea insecticides in infused oolong tea
  • Multi-residue analysis of pesticides in Açai fruit by an extraction based on matrix solid phase dispersion and determination using liquid chromatography
  • Comparative study for the extraction of 9 benzoylurea insecticides using two dispersive liquid-liquid microextraction (DLLME) approaches from wastewater samples followed by their determination using liquid chromatography-quadrupole-linear ion trap-mass spectrometry
Attention: P260 - P273 - P314 - P391 - P501
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité