Supelco® TraceCERT® Organic Reference Standard, Dichlofluanid
Fournisseur: Merck
CRM04195-100MG
CRM04195-100MGEA 77.2 EUR InStock
CRM04195-100MG
Supelco® TraceCERT® Organic Reference Standard, Dichlofluanid
Étalons Étalons organiques
It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Dichlofluanid CRM may also find its use as described below:
  • To determine the concentration of dichlofluanid and its metabolite in seawater and sediment of three marinas in Greece before and during the yachting season
  • To evaluate the biochemical and physiological effects of dichlofluanid exposure on the brown mussel Perna perna
  • Photocatalytic degradation of chlorothalonil and dichlofluanid over TiO₂ suspensions
  • Testing toxicity of the fungicides penconazol, cymoxanil, and dichlofluanid and their effects on the specific growth rate, biomass production, respiration rate, and yeast viability
  • To estimate the rates of disappearance of dichlofluanid and tebuconazole following successive applications to two varieties of lettuce grown in greenhouse conditions
Attention: P261 - P273 - P280 - P302 + P352 - P304 + P340 + P312 - P305 + P351 + P338
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité