Supelco® TraceCERT® Organic Reference Standard, Ethiofencarb
Fournisseur: Merck
CRM05286-100MG
CRM05286-100MGEA 77.8 EUR InStock
CRM05286-100MG
Supelco® TraceCERT® Organic Reference Standard, Ethiofencarb
Étalons Étalons organiques
The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. The CRM may also find the following uses:
  • Estimation of systemic and non-systemic pesticides in 80 samples of apples from Kazakhstan by LC-MS/MS and GC-MS/MS
  • Development of a method for rapid multi-residue screening of 12 pesticides and 7 mycotoxins in grain and animal feed samples using flow injection (FI) mass spectrometry (MS)
  • UHPLC-MS/MS determination of carbamate pesticide residues in fresh herb samples for human consumption and an optimised QuEChERS extraction
  • Study the photocatalytic degradation of ethiofencarb in the presence of a chalcogenide, SnI₄S₈
  • Differentiation between structurally isomeric pesticides using high-resolution accurate mass analyzers
  • Multi-residue analysis of 250 pesticides by ultra-performance liquid chromatography (UHPLC) coupled to MS/MS in processed fruits
Certifications: The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. The CRM may also find the following uses:
  • Estimation of systemic and non-systemic pesticides in 80 samples of apples from Kazakhstan by LC-MS/MS and GC-MS/MS
  • Development of a method for rapid multi-residue screening of 12 pesticides and 7 mycotoxins in grain and animal feed samples using flow injection (FI) mass spectrometry (MS)
  • UHPLC-MS/MS determination of carbamate pesticide residues in fresh herb samples for human consumption and an optimized QuEChERS extraction
  • Study the photocatalytic degradation of ethiofencarb in the presence of a chalcogenide SnI4S8
  • Differentiation between structurally isomeric pesticides using high-resolution accurate mass analyzers
  • Multi-residue analysis of 250 pesticides by ultra-performance liquid chromatography (UHPLC) coupled to MS/MS in processed fruits
Attention: P264 - P270 - P273 - P301 + P312 - P391 - P501
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité