Supelco® TraceCERT® Organic Reference Standard, Anilazine
Fournisseur: Merck
CRM05691-25MG
CRM05691-25MGEA 42.1 EUR InStock
CRM05691-25MG
Supelco® TraceCERT® Organic Reference Standard, Anilazine
Étalons Étalons organiques
The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. The CRM may also find the following uses:
  • Analysis of long-term persistence of three 14C-labeled pesticides- ethidimuron, methabenzthiazuron, anilazine, and their metabolites in soil
  • Gas chromatographic-mass spectrometric (GC-MS) determination of different classes of pesticide residues from various wastewater samples obtained from culturing and processing of potatoes
Attention: P264 - P273 - P280 - P302 + P352 - P305 + P351 + P338 - P332 + P313
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité