Mouse Reactive Inflammasome Antibody Sampler Kit
Fournisseur: Cell Signaling
20836T
CSTA20836TEA 668 EUR InStock
CSTA20836T
Mouse Reactive Inflammasome Antibody Sampler Kit
Anticorps
The Mouse Reactive Inflammasome Antibody Sampler Kit provides an economical means of detecting multiple inflammasome components. The kit includes enough antibodies to perform at least two western blot experiments with each primary antibody.
Each antibody in the Mouse Reactive Inflammasome Antibody Sampler Kit detects endogenous levels of its target protein. Caspase-1 (E2Z1C) Rabbit mAb detects endogenous levels of full-length mouse caspase-1; this antibody detects pro-caspase-1 and the p10 subunit of activated caspase-1. Cleaved Caspase-1 (Asp296) (E2G2I) Rabbit mAb detects endogenous levels of mouse caspase-1 only when cleaved at Asp296. A non-specific band is detected at 70 kDa in some cells. Cleaved-IL-1β (Asp117) (E7V2A) Rabbit mAb (Mouse Specific) recognises endogenous levels of mouse IL-1β protein only when cleaved at Asp117.
Information de livraison: Kit includes NLRP3 (D4D8T) Rabbit mAb (CSTA15101) 20 µl, AIM2 Antibody (CSTA63660) 20 µl, ASC/TMS1 (D2W8U) Rabbit mAb (CSTA67824) 20 µl, Cleaved-IL-1β (Asp117) (E7V2A) Rabbit mAb (CSTA63124) 20 µl, IL-1β (D6D6T) Rabbit mAb (CSTA31202) 20 µl, Cleaved Caspase-1 (Asp296) (E2G2I) Rabbit mAb (CSTA89332) 20 µl, Caspase-1 (E2Z1C) Rabbit mAb (CSTA24232) 20 µl, Anti-rabbit IgG 20 µl, HRP-linked Antibody (CSTA7074) 100 µl.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité