Cryostat, LS3000-A
Fournisseur: DAKEWE MEDICAL
130.012.007-00
DAKE130.012.007-00EA 0 EUR InStock
DAKE130.012.007-00
Cryostat, LS3000-A
Microtomes et groupes de microtomes Microtomes
Cryostat has a injection molding shell with mild feeling operation, durable and easy to clean and also has four colours of specimen chucks contribute a convenient management of specimens.
  • Adjustable cool plate temperature with −45 °C
  • Super large, may hold 28 chucks simultaneously
  • Adjustable peltier temperature, minimum temperature is −65 °C
  • Cleaning system
  • Universal hand wheel can be locked at any position
  • 40 db silent freezing design with dual compressors from Germany
  • Anti-fog glass window
New designed hammer make the tissue platter than the traditional one with no condensation.Blade holder auto pump-blade device avoids incised wound.
Dimensions: 700×660×1250 mm
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité