Sleeve protector, cut resistant, Armcut 14 BT
Fournisseur: HONEYWELL SAFETY
4150057
DALL4150057EA 17.5 EUR InStock
DALL4150057
Sleeve protector, cut resistant, Armcut 14 BT
Tenue de travail
This 2-ply Kevlar® thread knitted sleeve provides protection of the forearm against cuts.
  • Tip of the sleeve stitched like a mitten
  • With thumbhole
  • Kevlar® fibre offers good cutting resistance and protection against thermal hazards (contact with heat)
  • Doubly thick, the cuff's mechanical and thermal performance is greater
  • Length of 36 cm ensures vast protection of the forearm
Typical applications include: Automotive and part manufacturing, building and construction, glass industries, iron and steel industry, paper industries, ship building, wood industries.
Gauge: 10
Certifications: EN 388 (132X), EN 407 (X1XXXX); Cat. II
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité