Sleeve protectors, cut resistant, Aracut®
Fournisseur: HONEYWELL SAFETY
4402835
DALL4402835EA 34.7 EUR InStock
DALL4402835
Sleeve protectors, cut resistant, Aracut®
Tenue de travail
Knitted, double Kevlar® sleeve with thumb hole, to protect the underarm and palm against cuts.
  • Close-fitting sleeve with good tactile sensitivity, optimum wearer comfort and increased strength
  • Kevlar® fibre offers excellent cut resistance and protects against thermal risks (contact heat)
  • Double thickness increases the mechanical and thermal resistance
  • Length guarantees full protection of the forearm
Ideal for use in the glass, iron and steel industry, in metalworking, in the paper and wood industry, in shipbuilding and in the construction industry.
Gauge: 10
Certifications: EN 407 (X1XXXX), EN 388 (132X), Cat. II
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité