Powered air purifying respirator (PAPR) kit, Compact Air® 200
Fournisseur: HONEYWELL SAFETY
A150401
DALLA150401EA 0 EUR InStock
DALLA150401
Powered air purifying respirator (PAPR) kit, Compact Air® 200
Respirators Systèmes de respiration
The Compact Air® 200 powered air supply system is designed to provide respiratory protection against gases, vapour and particles depending on the type of filter element used.
  • Impact resistant ABS blower unit
  • In-use and storage temperature range of –10 to +50 °C
Particulates only and combined particulates, gas and vapour. Lithium-ion rechargeable battery pack and housing has a operating time of 8 - 13 hours.
Certifications: EN 12941 (TH1-TH2-TH3) / EN 12942* (TM2-TM3) / CE 0194
Information de livraison: Kit includes blower unit, lithium-ion battery pack and housing, universal battery charger, belt with comfort pad, breathing tube for hoods (840 mm length), air flow indicator and polypropylene carrying case with moulded foam lining.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité