Perfect Fit™ Cut Resistant Gloves
Fournisseur: HONEYWELL SAFETY
NPF21-1118W-11
DALLNPF21-1118W-11EA 0 EUR InStock
DALLNPF21-1118W-11
Perfect Fit™ Cut Resistant Gloves
Gants
The Honeywell Perfect Fit™ gloves were created with comfort in mind while also giving premium protection. The Perfect Fit™ Gloves are designed to protect workers from cuts, abrasions, lacerations, and punctures while working around sharp and rough materials.
  • Low content level of dmf (<100)
  • Colour coded markings
  • Silicon-free option
  • Coating breathability
  • Glass-fiber free for comfort
  • Light weight and Washable (up to 5 cycles of washing)
  • High grip performance
Perfect Fit™ has the unique Honeywell colour coding system for ease of identification, we also equipped this glove with a QR code to quickly give users access to technical information on the different cut levels and silicon free options.
Used industries are automotive, machinery and equipment, metal fabrication, warehousing and logistics, manufacturing, construction, transportation.
Applications: Handling sharp objects, painting and surface inspections, chassis assembly, mounting and dismantling, maintenance, fastening components and parts.
Information de livraison: Each packs contains 10 pairs.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité