Temperature and humidity loggers, Compact
Fournisseur: DICKSON
SP125
DICKSP125EA 0 EUR InStock
DICKSP125
Temperature and humidity loggers, Compact
Enregistreurs de données
The data loggers is self-contained with high speed USB connectivity, high/low LED alarm, an internal sensor for recording air temperatures and a replaceable battery for accurate, easy-to-use and PC-based recording. The polycarbonate casing, compact size and rugged design allows for use in numerous applications like regulatory compliance, HVAC balancing, warehouse mapping, process control.
Certifications: IP 20 certified protection class.
Information de livraison: Supplied with CR24450 lithium battery, product comes with one year of warranty.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité