Converts Liquid waste to solid waste for easy disposal. Super absorbent granules can rapidly absorb aqueous or mixed aqueous and organic solvent waste. Common uses: Put Lab-Sorb at the bottom of biohazard bin, Liquid containment for spills, convert 1 * 1 Pce
Fournisseur: DIVERSIFIED BIOTECH
Converts Liquid waste to solid waste for easy disposal. Super absorbent granules can rapidly absorb aqueous or mixed aqueous and organic solvent waste. Common uses: Put Lab-Sorb at the bottom of biohazard bin, Liquid containment for spills, convert 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Converts Liquid waste to solid waste for easy disposal.  Super absorbent granules can rapidly absorb aqueous or mixed aqueous and organic solvent waste.  Common uses: Put Lab-Sorb at the bottom of biohazard bin, Liquid containment for spills, convert 1 * 1 Pce DIVESORB-1000 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou