Soft Care Des E Alcohol Based Hand Disinfectant
Fournisseur: Diversey
100962439
DIVY100962439EA 199 EUR InStock
DIVY100962439
Soft Care Des E Alcohol Based Hand Disinfectant
Distributeurs de savon et distributeurs de savon liquide
Soft Care Des E is an alcohol based 71,5% hand disinfectant gel for waterless skin disinfection. Soft Care Des E is suitable for use in all areas, including healthcare and food processing environments.
  • Contains ethanol, a highly effective disinfectant
  • Suitable for surgical disinfection
  • Fragrance and colour-free; Suitable for use in the food processing industry
  • Contains an effective moisturiser preventing skin irritation and dehydration
Hand disinfectant based on ethanol. Ethanol is a disinfectant which is very effective against resident and transient flora such as bacteria (e.g. MRSA) and yeast. It is also effective against enveloped viruses such as influenza and certain non-enveloped viruses such as Norovirus and Adenovirus.
Perfume-free eliminating the risk of tainting foods and making it suitable for application in food processing areas. Skin compatibility dermatologically tested (Patch and TEWL tests). Contains a skin softener helping the skin to remain smooth and intact, particularly in areas where frequent hand disinfection is required.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité