Oxivir Excel® Cleaning and Disinfectant Wipes
Fournisseur: Diversey
100984249
DIVY100984249EA 144 EUR InStock
DIVY100984249
Oxivir Excel® Cleaning and Disinfectant Wipes
Lingettes Lingettes et essuie-mains à usage unique Lingettes désinfectantes
Diversey Oxivir Excel® combined cleaner and disinfectant, powered by AHP technology, provides you with the best alternative to your traditional disinfectants by delivering afull virucidal efficacy in just 30 seconds, while being very gentle to surfaces and withthe lowest risk to your staff and customer. The active ingredient, hydrogen peroxide,breaks down into environmentally-friendly water and oxygen just minutes after useleaving no residues that might be harmful or change the appearance of the surface.
  • Full virucidal claim in just 30 s
  • Fast kill claims for a broad spectrum of viruses,
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité