ClearKlens Solve VH21 Detergent
Fournisseur: Diversey
101103248
DIVY101103248EA 137 EUR InStock
DIVY101103248
ClearKlens Solve VH21 Detergent
Produits nettoyants Détergents et Nettoyants de Surfaces
ClearKlens solve is an alkaline foam detergent for use in the pharmaceutical and cosmetic industries.
  • Balanced blend of anionic and non-ionic surfactants in alkaline solution
  • Removes oils, fats and grease of vegetable and animal origin and other product residues found in the pharmaceutical and cosmetic industries
  • ClearKlens solve is for manual cleaning of process equipment, floors and walls and foam applications
  • Highly effective alkaline detergent for difficult cleaning tasks
  • Can be used on aluminum with care as directed
Use ClearKlens Solve for dailyMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité