TASKI® Sprint 200 Pur-Eco E1b Multi Purpose Cleaner
Fournisseur: Diversey
7515239
DIVY7515239EA 28.4 EUR InStock
DIVY7515239
TASKI® Sprint 200 Pur-Eco E1b Multi Purpose Cleaner
Produits nettoyants Détergents et Nettoyants de Surfaces
Alcohol-based daily multi purpose cleaner for streak-free cleaning of all water resistant hard surfaces.
  • pH neutral at in use dilution
  • Fast acting, fast drying
  • Based on an effective combination of alcohol and surfactants derived from natural vegetable sources to minimise the impact on the environment and ensure safer use
  • Unified fresh fragrance throughout the Pur-Eco range
Benefits: Suitable for use on water-resistant hard surfaces, including windows, mirrors, glazed ceramics as well as plastic surfaces. Leaves surfaces free of marks and streaks. Excellent removal of fingermarks as well the spot treatment of other slightly adherent soils. Leaves a pleasant lime scent in all cleaned areas.
Certifications: EU Ecolabel.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité