Insulated boxes, TransPorter Blueline
Fournisseur: DELTA T
B100020DE B100031 B100034 B100050 B100023 B100076
DLTTB100020DEEA 272 EUR InStock
DLTTB100020DE DLTTB100031 DLTTB100034 DLTTB100050 DLTTB100023 DLTTB100076
Insulated boxes, TransPorter Blueline
Boîtes Boîtes de transport
TransPorter Blueline boxes are robust, stackable, dishwasher safe and sealable.
  • With ergonomic handles
  • The box is not easy to be damaged
  • Multiple boxes can be stacked for transportation
  • Suitable for TempShell system
  • A seal can be used to prevent unauthorised opening of the transport containers
Optional inclusion of RFID-chip for temperature monitoring and box identification (enquire for details).
Thermal insulation box with thick thermal insulation material. Vacuum insulation and transparent plastic inliner with cavities for cooling elements. There are three different positions in the box fit S 1 L elements, and at the same time, two 1 L elements can be placed at the bottom according to actual needs.
Commander maintenant
COMMANDE
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité