Molykote® High Vaccum Grease
Fournisseur: DOW CORNING
23900246
DOWC23900246EA 49 EUR InStock
DOWC23900246
Molykote® High Vaccum Grease
Lubrifiants
Molykote® High-Vacuum grease features a stiff, non-melting silicone formula that helps to seal and lubricate chemical processing equipment. Suitable for use in a wide range of applications including sealing vacuum and pressure systems, it is also doubles as a lubricant for o-rings in binoculars and telescopes as it prevents fogging of delicate lenses.
  • Helps to reduce wear in plastic-to-metal and rubber-to-metal contact
  • Low volatility for applications involving chemical processing equipment
  • Excellent resistance to water, chemicals, high and low temperatures
  • Helps prevent fogging of delicate lenses
Applications: Lubrication, sealing, chemical processing equipment, O-rings, vaccum and pressure systems.
Certifications: Meets FDA 21 CFR 175.300 and NSF 61.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité