PAS® AirPack 1 Compressed Air Breathing Apparatus
Fournisseur: Dräger
3352228
DRAE3352228EA 0 EUR InStock
DRAE3352228
PAS® AirPack 1 Compressed Air Breathing Apparatus
Respirators Systèmes de respiration
This heavy duty airline apparatus is ideal for use where an extended duration of breathing air is called for. Chemical tank cleaning, toxic spillages or certain tasks when working on offshore installations are all made easier and more comfortable when using the PAS AirPack 1.
  • Increased freedom of movement
  • Anti-corrosion polymeric powder coating
  • Pneumatic arrangement
  • Hore drum with brake mechanism
  • Extendable hose upto 50 meter
  • Twin cylinder support straps
  • Trolley frame with two-wheeled fixed angle
A 50 metre length of hose is supplied with the AirPack 1 as standard, this can be extended to a length of 100 metres with a range of extension hoses. The trolley hose reel connector is protected with a cover to minimise inadvertent damage being caused.
Input pressure cylinder: 200 or 300 bar
Input pressure airline: 6 to 9 bar
Airline whistle activation pressure: 4 to 5 bar
Whistle sound level: >90 dBA
Operating temperature range: −32 to +70 °C
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité