X-am® 2500 Ex Gas Detection System
Fournisseur: Dräger
8323912
DRAE8323912EA 1130 EUR InStock
DRAE8323912
X-am® 2500 Ex Gas Detection System
Systèmes de détection de gaz
The X-am® 2500 was especially developed for use as personal protection. The 1 to 4 gas detector reliably detects combustible gases and vapours, as well as O₂, CO, H₂S, NO₂ and SO₂. Reliable and fully mature measuring technology, durable sensors and easy handling ensure a high degree of safety with extremely low operating costs.
  • Durable electrochemical sensors and poison resistant Ex sensor
  • Fast and easy maintenance, inexpensive
  • Ergonomic and robust
  • Reliable power supply
  • Pressure 700 to 1300 mbar
X-am 2500 system has Ex approval for zone 0, so it is clearly designed for very high user safety in areas subject to explosion hazard. The functional design ensures that gas can enter from above and from the front.
Thanks to its low weight and ergonomic design, the X-am 2500 offers a high degree of wearing comfort. The practical two button control panel and easy menu navigation allow the instrument to be used intuitively, despite its comprehensive functionality. The integrated protective rubber coating and sensors that are not sensitive to shock provide additional safety in case of impacts or vibrations.
Moreover, the X-am 2500 is not sensitive to electromagnetic radiation, e.g. from wireless devices. The X-am 2500 is water and dust resistant in accordance with protection class IP 67, so it remains fully functional even if it falls accidentally into the water.
The X-am 2500 can operate with either alkaline batteries or with rechargeable NiMH batteries. This enables a reliable power supply for more than 12 hours. Depending on the requirements, the batteries can be charged either in the workshop or in a vehicle. Operating time without Ex sensor is typically more than 250 hours.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité