Escape hoods, Parat® 5520
Fournisseur: Dräger
R59425
DRAER59425EA 306 EUR InStock
DRAER59425
Escape hoods, Parat® 5520
Respirators Systèmes de respiration
This combined fire and industrial escape hood was developed with users, placing the focus on the fastest possible escape. Optimised operation and wearing comfort, a robust housing and a tested CO P2 filter ensure protection from toxic fire-related gases, vapours and particles for at least 15 minutes while escaping.
  • Hood in signal colour for high visibility
  • Window enables you to check the expiration date and the device condition
  • Escape hood can be carried with a belt, shoulder strap, grip clip or belt clip
When opening the packaging, the filter plug is automatically released from the filter. The filter is then deployed into operational position and the hood can be immediately donned. Thanks to the self-adjusting internal head harness, no additional adjustment is required. Users only have to open the packaging, remove and don the hood – and leave the danger zone.
Total service life: 16 years (provided the filter is exchanged after eight years)
Certifications: DIN 58647-7 (filter additionally tested in accordance with EN 14387:2004)
Conditionnement: The Soft Pack provides dust protection (IP5). It has a viewing window to check the filter expiration date and the condition of the device. The security seal on the packaing shows if the unit has been opened.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité