Safety storage cabinets for toxic substances
Fournisseur: DUPERTHAL
20-201045-011
DUEH20-201045-011EA 1095.05 EUR InStock
DUEH20-201045-011
Safety storage cabinets for toxic substances
Mobilier Armoires de stockage
Safety storage cabinet ideal for storage of non flammable substances.
  • Torsion resistant wing doors with box gusset
  • Sheet steel cabinet with light grey colour RAL 7035
  • High quality safety lock for increased protection against break-ins
  • Four adjustable feet to compensate for uneven ground
  • Height adjustable perforated shelves for optimal ventilation
Cabinet features include wing door classical technology, integrated bracket for poison book with depth of 15 mm, bottom tray galvanised with material thickness 3 mm and ventilation via connection socket (NW 75/2,95) on the cabinet roof.
It is approved for storage of maximum 200 kg of class T toxic substances and maximum 50 kg of T+ highly toxic substances.
Certifications: Complies with requirements of German Equipment and Product Safety Act. The bottom tray is manufactured with declaration of conformity in accordance with StawaR and with Water Resources Act.
Attention: It is not suitable for storage of flammable liquids.
Commander maintenant
COMMANDE
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité