Cleanroom hoods with integrated mask, Tyvek® IsoClean®, Model IC689B WS
Fournisseur: DuPont
IC689BWHWS
DUPOIC689BWHWSEA 440 EUR InStock
DUPOIC689BWHWS
Cleanroom hoods with integrated mask, Tyvek® IsoClean®, Model IC689B WS
Vêtements à usage unique
These hoods with integrated mask, model IC689BWH WS, are made of Tyvek®, which is a fabric composed of flash spun high density polyethylene, providing an ideal balance of protection, durability, and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
  • Aseptically folded
  • Ties for adjustable fit in the chest area
  • Gamma irradiated to Sterility Assurance Level (SAL) of 10⁻⁶
  • Suitable for use in GMP class A/B (ISO Class 5) cleanrooms
The white hood features bound, internal seams and a face opening. Integrated mask is blue pleated polyethylene, measuring 175 mm wide. Note, mask is not a PPE garment (not FFP1, 2, or 3 mask).
All DuPont™ Tyvek® IsoClean® sterile accessories (option code WS) are packed in a dual barrier packaging system, consisting of an inner and outer easy-tear cleanroom bag.
Garments and accessories made of sterile Tyvek® IsoClean® are typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry, as well as in other critical or controlled environments.
Certifications: Partial body chemical protective clothing, Type PB [6-B], EN 14126, Cat. III
Conditionnement: 100 per box, individually double packed. Two polyethylene liners. Cardboard box.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité