Winlog.pro Evaluation Software for Ebro Dataloggers
Fournisseur: EBRO
1340-2450
EBRO1340-2450EA 1190 EUR InStock
EBRO1340-2450
Winlog.pro Evaluation Software for Ebro Dataloggers
Logiciels Logiciels de laboratoire
The easy-to-use software products are suitable for programming and read-out of Ebro® data loggers as well as for the evaluation of the measured values. For professional software.
  • Contains all properties of Winlog.basic
  • Enables real-time monitoring with radio wave data loggers
  • Current version: 3.1
Supports: EBI 12, EBI 10, EBI 11, EBI 20, EBI 40, EBI 100, EBI 300, EBI 310, EBI 330. Integration of images and diagrams into reports possible. FDA 21 CFR Part 11 functionality also possible. Optional IQ-/OQ documentation also available. Formula editor for calculating the F₀ value, the absolute humidity, the PE value, etc.
Hardware requirements: Processor - pentium 1 GHz, main memory - 1 GB, free hard disk space - 1 GB, USB.
Software requirements: Windows 8 (32 bit or 64 bit), windows 10 (32 bit or 64 bit).
Information de livraison: Includes calibration tool for the data logger.
Commander maintenant
COMMANDE
ACCESSOIRES
PRODUITS COMPLEMENTAIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité