Winlog.Validation Evaluation Software for Ebro Data Loggers EBI 12, EBI 11 and EBI 16
Fournisseur: EBRO
1340-2470
EBRO1340-2470EA 4220 EUR InStock
EBRO1340-2470
Winlog.Validation Evaluation Software for Ebro Data Loggers EBI 12, EBI 11 and EBI 16
Logiciels Logiciels de laboratoire
The easy-to-use software products are suitable for programming and read-out of Ebro® data loggers as well as for the evaluation of the measured values. For routine checks and validation.
  • User-friendly
  • Highly accurate measurements
  • Automatic report generation
  • Automatic user-defined calculations
  • Automatic recognition of process cycles
  • Current version: 4.0
Save time and effort for the regularly required process validation with the help of the powerful reporting and evaluation software Winlog.validation. The automatic recognition of the individual process steps makes your work easier and meets the high demands of validation and qualification in pharmacy and medicine.
High-performance report and evaluation software for the stringent requirements of validation and qualification in the pharmaceutical and medical sectors. Supports: EBI 12, EBI 10, EBI 11, EBI 16, EBI 20, EBI 40, EBI 100, EBI 300, EBI 310.
Creation of user-defined templates for specific devices and thermal processes. Three-dimensional display of sensor placement. FDA 21. CFR Part 11 functionality also possible. IQ-/OQ documentation available. Automatic completion of validation processes.
Hardware requirements: Processor speed minimum 1,6 GHz, working memory 2 GB (32 bit), working memory 4 GB (64 bit) 4 GB free hard disc space, USB A (universal serial bus).
Software requirements: Windows 8 (32 bit or 64 bit), windows 10 (32 bit or 64 bit), windows 11.
Commander maintenant
COMMANDE
PRODUITS COMPLEMENTAIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité