Winlog.Web Server Based Monitoring Software for EBI 25
Fournisseur: EBRO
1340-2490 1340-2491
EBRO1340-2490EA 1280 EUR InStock
EBRO1340-2490 EBRO1340-2491
Winlog.Web Server Based Monitoring Software for EBI 25
Logiciels Logiciels de laboratoire
The easy-to-use software products are suitable for programming and read-out of Ebro® data loggers as well as for the evaluation of the measured values. Supports: data loggers EBI 25 series, MODBUS IP devices. Professional version for multiple users.
  • Connection of the interface IF 400 via USB and ethernet
  • FDA 21 CFR Part 11 functionality
  • Administration of large quantities of data
  • IQ-/OQ documentation available
  • Current version: 3.11
Web-based server solution: the measurement data can be read on all computers and smartphones which are connected with theMore Product Information
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
PRODUITS COMPLEMENTAIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité