Set for Validation of Large Steam Sterilizers, SL 3101
Fournisseur: EBRO
1340-6024 1250-3101
EBRO1340-6024EA 287 EUR InStock
EBRO1340-6024 EBRO1250-3101
Set for Validation of Large Steam Sterilizers, SL 3101
Enregistreurs de données
The SL3101 set by ebro, featuring five EBI 12-T471 and one EBI 12-TP453 loggers in silicone protection boxes, is ideal for validating large sterilisers and autoclaves in demanding environments.
  • Comprehensive set for large sterilizers and autoclaves
  • Includes 5× EBI 12-T471 and 1× EBI 12-TP453 loggers
  • Silicone protection boxes (AL 107 and AL 101) for added heat resistans
  • Traceable factory calibration certificate
The SL3101 set by ebro is designed for the validation and monitoring of large sterilizers and autoclaves, essential for maintaining highMore Product Information
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
PRODUITS COMPLEMENTAIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité