ECHO Revolve 3 Hybrid Microscope (Upright + Inverted)
Fournisseur: DISCOVER ECHO
RVSB1000
ECHLRVSB1000EA 23970 EUR InStock
ECHLRVSB1000
ECHO Revolve 3 Hybrid Microscope (Upright + Inverted)
Microscopes
This 'all in one' hybrid microscope can be used for upright and inverted observation methods.
  • Intelligent objective nosepiece - this allows the software interface to automatically recognise the objective being used
  • Illumination technique: brightfield (phase contrast available as an option)
  • 5MP CMOS color camera
  • Touchscreen functions drive camera and illumination, replacing manual turrets and levers
  • Traditional eyepieces are replaced by an iPad® tablet for easier viewing (retina display)
  • Custom software app simplifies the workflow of taking images and seamlessly integrates with other cloud sharing apps
The upright observation method is ideal for viewing glass slides. Useful for water dipping applications.
The inverted observation method is suitable for viewing live samples in fluidic chambers such as dishes and well plates. It also allows for easier manipulation of samples. Ideal for long working distance applications.
Informations de commande: To be completed with objectives, which are sold separately.
Information de livraison: Includes an iPad® interface, power supply, dust cover, dark box, and lens tissue.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité