Tissu -,,HTS 0600 R 01500 W-5650 Highextra smooth hygienic, low linting, non-scratching, dry & wet use, very strong, no binders,resists to acids & solvents, good absorbency, silicon free 55% cellulose 45% polyester white: 68 gsm 1 * 5 ROU
Fournisseur: ECOLINE NV SA
Tissu -,,HTS 0600 R 01500 W-5650 Highextra smooth hygienic, low linting, non-scratching, dry & wet use, very strong, no binders,resists to acids & solvents, good absorbency, silicon free 55% cellulose 45% polyester white: 68 gsm 1 * 5 ROU
New Product
COMMANDE
Développer tout / Réduire tout
Tissu -,,HTS 0600 R 01500 W-5650 Highextra smooth hygienic, low linting, non-scratching, dry & wet use, very strong, no binders,resists to acids & solvents, good absorbency, silicon free 55% cellulose – 45% polyester white: ± 68 gsm 1 * 5 ROU ECOLHTS0600R01500W 1 * 5 ROU
New Product
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou