Nucleic acid testing for COVID-19
Fournisseur: EDVOTEK
123
EDVO123EA 139 EUR InStock
EDVO123
Nucleic acid testing for COVID-19
Matériel éducatif Matériel éducatif de biologie Activités de génétique
Hands-on simulation of molecular testing for COVID-19.
  • Students learn about characteristics of Coronaviruses
  • Includes all required reagents
  • Use DNA Electrophoresis to analyse simulated samples
  • Activity can be completed in 45 minutes
SARS-CoV-2 is a novel coronavirus that has caused a worldwide outbreak of respiratory disease beginning in 2019.
In this simulated medical test, we will use electrophoresis to detect the presence of the SARS-CoV-2 virus in samples from patients with symptoms of COVID-19.
Additional items needed but not included: DNA electrophoresis equipment, white-light box, microwave or hot plate.
Informations de commande: Kit will ship immediately upon ordering. Please call to specify a future shipping date.
Attention: Store DNA samples in the refrigerator. All other components may be stored at room temperature.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité