Using CRISPR to treat cystic fibrosis
Fournisseur: EDVOTEK
135
EDVO135EA 120 EUR InStock
EDVO135
Using CRISPR to treat cystic fibrosis
Matériel éducatif Matériel éducatif de biologie Activités de génétique
Hands-on simulation of CRISPR-Cas9 to target and treat Cystic Fibrosis.
  • Teach your students about new CRISPR technology
  • Includes all required reagents
  • Use DNA electrophoresis to view reach a diagnosis.
  • Activity can be completed in 45 minutes
In this experiment, students will simulate the use of CRISPR-Cas9 to target a genetic mutation found in a patient suffering from Cystic Fibrosis.
Students will develop an understanding of guide RNA (gRNA) design, and use agarose gel electrophoresis to examine pre-prepared DNA samples after CRISPR treatment.
Additional items needed but not included: DNA electrophoresis equipment, white-light box, microwave or hot plate.
Informations de commande: Kit will ship immediately upon ordering. Please call to specify a future shipping date.
Attention: Store DNA samples in the refrigerator. All other components may be stored at room temperature.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité