M12 Ultra™ Electrophoresis and Visualisation Package
Fournisseur: EDVOTEK
505
EDVO505EA 373 EUR InStock
EDVO505
M12 Ultra™ Electrophoresis and Visualisation Package
Systèmes d'électrophorèse Cuves d'électrophorèse horizontale
Our best-selling M12 Complete™ just got even better. The M12 Ultra™ includes the M12 Complete™ electrophoresis package and the all-new TruBlu™ Jr. Blue light transilluminator. This dynamic duo enables real-time visualisation of SYBR® Safe or other blue light-reactive dye agarose gels.
  • For blue light-reactive gels, overlay the orange filter on the M12 lid and slide the TruBlu™ Jr. underneath for instant visualisation
  • The M12 Ultra™ offers the ultimate flexibility to either visualize gels in real-time
  • The TruBlu™ Jr. can also be used to examine fluorescence from green fluorescent protein (GFP)
  • Produces excellent results in 15 to 30 minutes
TruBlu™ Jr. Features:
Battery operated for convenient use in the classroom
Designed to work seamlessly with the EDVOTEK® M12 Electrophoresis System
Includes an orange contrast lid
Safe, non-UV wavelength
Blue Light emission spectrum centered around 470nm is compatible with SYBR® Safe, GelGreen® & other 10×6 cm viewing surface
M12 Complete™ Features:
Sleek new design speeds electrophoresis
Includes complete set of new and improved electrophoresis accessories
Now includes a DNA DuraGel™ (Cat. #S-43) for pipetting practice
Large color coded push tabs for easy lid insertion and removal
Pour spout for buffer disposal
Improved ventilation reduces lid condensation
User replaceable electrodes
Chamber reverse compatible with previous Edvotek® accessories
Information de livraison: M12 Ultra™ Package includes: 2× Gel trays (7×7 cm), 1× gel tray (14×7 cm), 2× ⁶/₈ tooth combs, 4× rubber end caps, 1× DNA DuraGel™ (Cat. #S-43), 1× TruBlu™ Jr., 1× orange contrast lid.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité