Linking food science to biotechnology kit
Fournisseur: EDVOTEK
S-47
EDVOS-47EA 94.9 EUR InStock
EDVOS-47
Linking food science to biotechnology kit
Matériel éducatif Matériel éducatif de biologie Activités de biochimie
Investigate how agarose gel electrophoresis unlocks the color code used by food scientists to make colorful candies.
  • NGSS-aligned with MS-PS1
  • For 10 lab groups
  • Complete in 45 minutes
  • Explore the biochemistry behind colorful candies
  • Understand the biology behind gel electrophoresis
  • Perform agarose gel electrophoresis to separate different sized dye molecules
  • Extract, identify, and size different food dyes from popular candies
Students will extract color activities from common candies and separate the dyes on agarose gel electrophoresis. A fun lab extension involves the use of candy to build a DNA model.
Informations de commande: DNA electrophoresis, micropipettes: 5-50 µl (optional), white light box (Optional), and microwave or hot plate are not included.
Information de livraison: Kit includes instructions, Ready-to-Load QuickStrip™ dye samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), practice gel loading solution, disposable pipets, and microcentrifuge tubes.
Conditionnement: Room temperature storage.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité