High Performance Compact Pirani Gauge, APG200
Fournisseur: EDWARDS HIGH VACUUM METTLER
D1G1011100
EDWAD1G1011100EA 1080 EUR InStock
EDWAD1G1011100
High Performance Compact Pirani Gauge, APG200
Pompe Jauges de vide
Edwards new APG200 series high performance compact pirani gauge is the perfect vacuum measurement solution across a spectrum of applications due to its compact size, LED light ring, integrated set points and flexibility of connections/outputs. 25% reduction in volume compared to the APG100, without any loss in performance. Maximum cable length of 100 m
  • Light ring pressure indication, compact size, long filament
  • Integrated filter
  • Wide range power supply
  • Drop in compatible
  • Set point relay
  • 360 bright LED ring
Digital gauges sit in the same footprint as analogue gauges, making it easy for you to upgrade at a later date
should more data-collection/control be needed. 15 to 48 V power input is the widest available on the market, enabling you to run these gauges on whatever power. It is used in analytical instruments, semiconductor, medical.
Storage temperature of −30 to +70 °C. Single push button control. Labview drivers software.
Certifications: UKCA ceritified.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité