TIC instrument controller, 3 head
Fournisseur: EDWARDS HIGH VACUUM METTLER
D39700000
EDWAD39700000EA 0 EUR InStock
EDWAD39700000
TIC instrument controller, 3 head
Pompe Jauges de vide
TIC Instrument controller is a compact control with a large, clear graphical display, an intuitive user interface and serial communications.
  • Automatically recognises and controls active gauges, including APG100 Pirani, convection, thermocouple, strain, inverted magnetron, wide range and active ion gauges
  • Enables complete integration into PC and PLC controlled processes, all TIC variants include RS232 and RS485 interface
Windows™-based PC program provides full remote setup, control and data logging functions via the RS232 interface.
Informations de commande: Accessories are sold separately. Contact your Avantor representative for more information.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité