DNA 20 kb Plasmid and Linear Kit
Fournisseur: EKSIGENT (A PART OF AB SCIEX)
5311708
EKSI5311708EA 3340 EUR InStock
EKSI5311708
DNA 20 kb Plasmid and Linear Kit
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
The DNA 20 kb plasmid and linear kit is used to do isoform and purity analysis of plasmid and linearised dsDNA.
This kit supplies high analytical resolution and can analyse a wide range of linear double-stranded DNA fragments or plasmid isoforms. Plasmid isoforms from 2,000 bp to 19,000 bp can be separated, including super coiled, linear, and open circular species. The kit can do size estimation and purity analysis for linear dsDNA samples from 100 bp to 20,000 bp.
Information de livraison: This kit provides reagent volumes for analysis of 200 samples and includes: Acid wash/regenerating solution (0,1 N HCl) (125 ml)- 1, CE grade water (140 ml)- 2, DNA 20 kb plasmid and linear conditioning solution (135 ml)- 1, DNA 20 kb plasmid and linear gel (135 ml)- 1, DNA 20 kb plasmid and linear sample buffer (55 ml)- 1, SYBR™ gold nucleic acid gel stain1 (120 µl)- 6, DNA 20 kb plasmid test mix (8 ng in 20 µl)- 6 1 SYBR™ is a trademark of the life technologies corporation. SYBR™ Gold nucleic acid gel stain is not available for resale.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité