Stirring/Heating Mantles, SHM-200
Fournisseur: Cole-Parmer
04641-05 04641-15 04641-25 04641-35 04641-45 04641-55
ELET04641-05EA 990 EUR InStock
ELET04641-05 ELET04641-15 ELET04641-25 ELET04641-35 ELET04641-45 ELET04641-55
Stirring/Heating Mantles, SHM-200
Appareils chauffants Chauffe-ballons
Provides powerful stirring speeds up to 520 min⁻¹ and heating up to 450 °C.
  • Bidirectional stirring with magnetic stirring bar auto capture function
  • Thermal insulating cartridge provides maximum heat transfer
  • Chemical resistant polypropylene housing is 'safe to the touch'
Stir samples while heating up to 450 °C (842 °F). These durable stirring/heating mantles feature a bidirectional magnetic stirrer with speeds up to 520 min⁻¹. An 'auto-recapture' function reactivates stirring if it is interrupted or lost for any reason.
A built-in power controller with dial markings from one to ten offer great repeatability. Two coloured lights indicate 'power on' and 'heat on'. The replaceable coiled heating element suspended within a thermal insulating cartridge provides maximum heat transfer and support.
Unique ventilation slots and ceramic fiber insulation keep the chemical-resistant polypropylene exterior cool even while in use. A safety earth (ground) screen and double fuses provide added safety.
Information de livraison: Supplied with 1× magnetic stirring bar and a power cord with UK plug.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité