Water purification and distribution systems, CENTRA® R 60, 120 and 200
Fournisseur: ELGA LabWater
CN200RDM1-230
ELGACN200RDM1-230EA 0 EUR InStock
ELGACN200RDM1-230
Water purification and distribution systems, CENTRA® R 60, 120 and 200
Purificateurs d'eau Systèmes d'eau pure et ultra-pure
The CENTRA-R 60, 120 and 200 offer complete water purification, storage, control and distribution systems delivering water which has been purified using reverse osmosis, UV photo-oxidization, optional deionization and 0,2 μm filtration.
  • Optional cartridge-based deionisation (simply plug and dispense)
  • Extended deionization performance with optional CO₂ degassing
  • UV photo-oxidation for enhanced bacterial control and TOC performance
  • Optional 0,2 µm filter for enhanced particle control
  • Composite vent filter helps maintain high water purity by preventing airborne gases and bacteria entering the reservoir
  • Access to a wide range of system monitoring and safety features via the central 'softkey' control panel
  • Additional safety monitoring, including product water quality alarm and consumable change reminder
R 60 and R 120 feature a flow rate of up to 10 liters/min and an integral 50 liter reservoir. R 200 features a loop flow rate up to 18 liters/min with standard system or up to 30 liters/min with the high flow rate variable speed model and an integral 350 liter reservoir.
Additional features of the R 200 system:
- Optional deionization (or other purification media) may be positioned outside the CENTRA-R 200, but within the loop
- 0,2 µm filter to maintain high water purity in the distribution loop
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité