Water storage and recirculating distribution system, CENTRA® RDS
Fournisseur: ELGA LabWater
CNXXXRDM1-230
ELGACNXXXRDM1-230EA 0 EUR InStock
ELGACNXXXRDM1-230
Water storage and recirculating distribution system, CENTRA® RDS
Purificateurs d'eau Systèmes d'eau pure et ultra-pure
The CENTRA-RDS is a storage and recirculating distribution system designed to work with other CENTRA or PURELAB systems or existing purification installations. In addition to storing and distributing pure water, the CENTRA-RDS maintains and improves water purity.
  • Delivers up to 30 L per minute of type I, type II and type III water from a distribution loop
  • Bacterial specification <5 CFU/ml is maintained using a combination of technologies
  • Reliable continual supply of pure water using unique access controls and leak detection system with optional building management system (BMS) connectivity
  • Designed to operate with other ELGA LabWater systems or existing purification installations
  • Compact design provides more flexibility for new buildings and refurbishments
  • Full service and validation support
The CENTRA RDS stores, controls and distributes pre-purified water throughout a building, or a suite of laboratories, via a loop system.
Information de livraison: Supplied with 0,2 μm filter, UV photo-oxidation, optional deionization connection, and a Central Storage and Distribution system with 350 L reservoir.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité